ENAH5802E141 Enamine

3-(1-amino-2-methylbutyl)benzoic acid hydrochloride

  • MDL number MFCD32068645
  • Cas Number : 2361743-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427762

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427762
Availability details for
Quantity
  • Availability
  • Add to Cart