ENAH5802E146 Enamine

4-(piperidin-1-yl)piperidine-4-carboxylic acid

  • MDL number MFCD11053550
  • Cas Number : 933721-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-5064752

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5064752
Availability details for
Quantity
  • Availability
  • Add to Cart