ENAH5802E148 Enamine

5-(aminomethyl)thieno[3,2-b]thiophene-2-carboxylic acid hydrochloride

  • MDL number MFCD32069141
  • Cas Number : 2375269-74-0
  • Molecular Weight : 249.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(aminomethyl)thieno[3,2-b]thiophene-2-carboxylic acid hydrochloride

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7438905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438905
Availability details for
Quantity
  • Availability
  • Add to Cart