ENAH5802E14D Enamine

2-(prop-2-en-1-ylsulfanyl)-4,5-dihydro-1,3-thiazole

  • MDL number MFCD00040453
  • Cas Number : 3571-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7226852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7226852
Availability details for
Quantity
  • Availability
  • Add to Cart