ENAH5802E14E Enamine

(2R)-2-amino-3-(5-chloro-2-fluorophenyl)propanoic acid hydrochloride

  • MDL number MFCD32198370
  • Cas Number : 2375248-37-4
  • Molecular Weight : 254.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7443103

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443103
Availability details for
Quantity
  • Availability
  • Add to Cart