ENAH5802E152 Enamine

2-amino-3-(7-methyl-1-benzofuran-3-yl)propanoic acid hydrochloride

  • MDL number MFCD32066295
  • Cas Number : 2361635-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7430403

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430403
Availability details for
Quantity
  • Availability
  • Add to Cart