ENAH5802E16F Enamine

6-amino-2-bromo-3-methoxybenzoic acid

  • MDL number MFCD15144685
  • Cas Number : 152946-38-8
  • Molecular Weight : 246.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-218557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-218557
Availability details for
Quantity
  • Availability
  • Add to Cart