ENAH5802E218 Enamine

(5-bromo-4-methyl-4H-1,2,4-triazol-3-yl)methanamine dihydrobromide

  • MDL number MFCD32198282
  • Cas Number : 2375273-92-8
  • Molecular Weight : 352.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HBr
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7442878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442878
Availability details for
Quantity
  • Availability
  • Add to Cart