ENAH5802E38F Enamine

methyl 7-bromo-1H-1,3-benzodiazole-2-carboxylate

  • MDL number MFCD28947549
  • Cas Number : 1803896-95-8
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-3563048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3563048
Availability details for
Quantity
  • Availability
  • Add to Cart