ENAH5802E393 Enamine

3-(4-bromophenyl)cyclobutane-1-carboxylic acid

  • MDL number MFCD26632936
  • Cas Number : 149506-16-1
  • Molecular Weight : 255.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-90
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-170614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170614
Availability details for
Quantity
  • Availability
  • Add to Cart