ENAH5802E3B3 Enamine

methyl 2-amino-3-(5-bromopyridin-3-yl)propanoate

  • MDL number MFCD24521294
  • Cas Number : 1565555-15-8
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6229858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6229858
Availability details for
Quantity
  • Availability
  • Add to Cart