ENAH5802E3B6 Enamine

7-iodo-1H-pyrazolo[4,3-b]pyridin-3-amine

  • MDL number MFCD22627840
  • Cas Number : 1350648-21-3
  • Molecular Weight : 260.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1689056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1689056
Availability details for
Quantity
  • Availability
  • Add to Cart