ENAH5802E3C5 Enamine

ethyl 5-bromo-2-ethyl-1,3-thiazole-4-carboxylate

  • MDL number MFCD32069285
  • Cas Number : 848555-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7429768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7429768
Availability details for
Quantity
  • Availability
  • Add to Cart