ENAH5802E3C6 Enamine

N-[(2-bromophenyl)methyl]methanesulfonamide

  • MDL number MFCD07056659
  • Cas Number : 889131-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7425170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7425170
Availability details for
Quantity
  • Availability
  • Add to Cart