ENAH5802E3C7 Enamine

ethyl 6-bromo-4-chloropyridine-3-carboxylate

  • MDL number MFCD28734542
  • Cas Number : 1335054-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7442302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442302
Availability details for
Quantity
  • Availability
  • Add to Cart