ENAH5802E3C9 Enamine

methyl 4-bromo-5-nitrothiophene-2-carboxylate

  • MDL number MFCD18905589
  • Cas Number : 31862-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-103
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2939623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2939623
Availability details for
Quantity
  • Availability
  • Add to Cart