ENAH5802E3CA Enamine

N-(5-bromo-4-methylpyridin-3-yl)aminosulfonamide

  • Cas Number : 2168761-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-3328680

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3328680
Availability details for
Quantity
  • Availability
  • Add to Cart