ENAH5802E3D1 Enamine

5-bromo-1-oxo-1,2-dihydroisoquinoline-3-carboxylic acid

  • MDL number MFCD32066440
  • Cas Number : 1597387-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-4480495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4480495
Availability details for
Quantity
  • Availability
  • Add to Cart