ENAH5802E3D2 Enamine

5-bromo-3-formyl-1H-indole-2-carboxylic acid

  • MDL number MFCD11107427
  • Cas Number : 1082193-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6502950

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6502950
Availability details for
Quantity
  • Availability
  • Add to Cart