ENAH5802E3D3 Enamine

6-bromo-3-formyl-1H-indole-2-carboxylic acid

  • MDL number MFCD07364678
  • Cas Number : 893730-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7430031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430031
Availability details for
Quantity
  • Availability
  • Add to Cart