ENAH5802E3D5 Enamine

methyl 5-bromo-2-methyl-1H-indole-3-carboxylate

  • MDL number MFCD22558437
  • Cas Number : 1225382-91-1
  • Molecular Weight : 268.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-bromo-2-methyl-1H-indole-3-carboxylate
Inchi Code 1S/C11H10BrNO2/c1-6-10(11(14)15-2)8-5-7(12)3-4-9(8)13-6/h3-5,13H,1-2H3
Inchi Key IMOLADHPXONWFR-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7430561

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7430561
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart