ENAH5802E3DA Enamine

2-(6-bromo-1-oxo-2,3-dihydro-1H-inden-2-yl)acetic acid

  • MDL number MFCD26749218
  • Cas Number : 181477-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7430538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430538
Availability details for
Quantity
  • Availability
  • Add to Cart