ENAH5802E3DC Enamine

2-(5-bromo-1-oxo-2,3-dihydro-1H-inden-2-yl)acetic acid

  • MDL number MFCD31384925
  • Cas Number : 69099-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174-176
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7440635

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440635
Availability details for
Quantity
  • Availability
  • Add to Cart