ENAH5802E3DD Enamine

6-bromo-3-chloro-2-fluoro-4-nitroaniline

  • MDL number MFCD32197987
  • Cas Number : 2375269-72-8
  • Molecular Weight : 269.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-3-chloro-2-fluoro-4-nitroaniline

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7441640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7441640
Availability details for
Quantity
  • Availability
  • Add to Cart