ENAH5802E3DF Enamine

ethyl 1-bromoimidazo[1,5-a]pyrazine-3-carboxylate

  • Molecular Weight : 270.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 1-bromoimidazo[1,5-a]pyrazine-3-carboxylate
Inchi Code 1S/C9H8BrN3O2/c1-2-15-9(14)8-12-7(10)6-5-11-3-4-13(6)8/h3-5H,2H2,1H3
Inchi Key AHUWOOAHQFAMAP-UHFFFAOYSA-N

Description

Country of Origin UA

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439124
Availability details for
Quantity
  • Availability
  • Add to Cart