ENAH5802E3E3 Enamine

2-{[(4-bromophenyl)methyl]amino}-N,N-dimethylacetamide hydrochloride

  • Cas Number : 2375271-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7441083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7441083
Availability details for
Quantity
  • Availability
  • Add to Cart