ENAH5802E3E5 Enamine

1-(5-chloro-4-iodo-1-methyl-1H-pyrazol-3-yl)methanamine hydrochloride

  • MDL number MFCD32069417
  • Cas Number : 2375268-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7436722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7436722
Availability details for
Quantity
  • Availability
  • Add to Cart