ENAH5802E3E6 Enamine

8-bromo-2-(methylsulfanyl)-3H,4H-pyrido[4,3-d]pyrimidin-4-one

  • MDL number MFCD30721047
  • Cas Number : 1709905-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7441418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7441418
Availability details for
Quantity
  • Availability
  • Add to Cart