ENAH5802E3E8 Enamine

5-chloro-3-fluoro-4-iodopyridin-2-amine

  • MDL number MFCD32065928
  • Cas Number : 1064783-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125738
Availability details for
Quantity
  • Availability
  • Add to Cart