ENAH5802E3EA Enamine

3-bromo-5-chloro-6-methoxyquinoline

  • MDL number MFCD29090868
  • Cas Number : 2092491-90-0
  • Molecular Weight : 272.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-5-chloro-6-methoxyquinoline
Inchi Code 1S/C10H7BrClNO/c1-14-9-3-2-8-7(10(9)12)4-6(11)5-13-8/h2-5H,1H3
Inchi Key QKEPLGZIKVJWNG-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7433129

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7433129
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart