ENAH5802E3F1 Enamine

(1R)-1-(2-bromo-4,5-dimethoxyphenyl)propan-1-amine hydrochloride

  • MDL number MFCD24415420
  • Cas Number : 2375247-64-4
  • Molecular Weight : 310.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-1-(2-bromo-4,5-dimethoxyphenyl)propan-1-amine hydrochloride
Inchi Code 1S/C11H16BrNO2.ClH/c1-4-9(13)7-5-10(14-2)11(15-3)6-8(7)12;/h5-6,9H,4,13H2,1-3H3;1H/t9-;/m1./s1
Inchi Key MYJYRENHSRKMRL-SBSPUUFOSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7439169

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7439169
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart