ENAH5802E3F2 Enamine

(1S)-1-(2-bromo-4,5-dimethoxyphenyl)propan-1-amine hydrochloride

  • MDL number MFCD24427440
  • Cas Number : 2375248-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7438434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438434
Availability details for
Quantity
  • Availability
  • Add to Cart