ENAH5802E3F4 Enamine

4-(4-bromo-2-chlorophenyl)piperidine hydrochloride

  • MDL number MFCD32065968
  • Cas Number : 2361636-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7431379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431379
Availability details for
Quantity
  • Availability
  • Add to Cart