ENAH5802E3F9 Enamine

2-bromo-5-chloro-1,3-bis(propan-2-yl)benzene

  • MDL number MFCD31693031
  • Cas Number : 2143028-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746602

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746602
Availability details for
Quantity
  • Availability
  • Add to Cart