ENAH5802E3FB Enamine

ethyl 5-bromo-2-cyclopropyl-1,3-thiazole-4-carboxylate

  • MDL number MFCD19703230
  • Cas Number : 1379345-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427702
Availability details for
Quantity
  • Availability
  • Add to Cart