ENAH5802E407 Enamine

2-bromo-4-methanesulfonylbenzoic acid

  • MDL number MFCD25966384
  • Cas Number : 7041-52-3
  • Molecular Weight : 279.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 208-210
Iupac Name 2-bromo-4-(methylsulfonyl)benzoic acid
Inchi Code 1S/C8H7BrO4S/c1-14(12,13)5-2-3-6(8(10)11)7(9)4-5/h2-4H,1H3,(H,10,11)
Inchi Key BAKURTCIMUEDPT-UHFFFAOYSA-N

Description

Melting Point 208-210
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-3375660

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-3375660
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart