ENAH5802E408 Enamine

1-(4-bromophenoxy)-4-methoxybenzene

  • MDL number MFCD18207480
  • Cas Number : 42203-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1709735

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1709735
Availability details for
Quantity
  • Availability
  • Add to Cart