ENAH5802E40E Enamine

7-bromo-1-(oxan-2-yl)-1H-indazole

  • MDL number MFCD22380250
  • Cas Number : 1158680-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434995
Availability details for
Quantity
  • Availability
  • Add to Cart