ENAH5802E415 Enamine

methyl 2-bromo-4-(difluoromethyl)-6-fluorobenzoate

  • Cas Number : 2092337-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7441976

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7441976
Availability details for
Quantity
  • Availability
  • Add to Cart