ENAH5802E418 Enamine

2,6-difluoro-4-iodobenzoic acid

  • MDL number MFCD11845965
  • Cas Number : 1160575-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1670104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1670104
Availability details for
Quantity
  • Availability
  • Add to Cart