ENAH5802E419 Enamine

sodium 2-{6-bromo-8-methoxyimidazo[1,2-a]pyridin-2-yl}acetate

  • Cas Number : 2375270-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7441322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7441322
Availability details for
Quantity
  • Availability
  • Add to Cart