ENAH5802E41A Enamine

lithium(1+) 4-(5-bromopyridin-2-yl)oxane-4-carboxylate

  • MDL number MFCD32066493
  • Cas Number : 2361635-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432124
Availability details for
Quantity
  • Availability
  • Add to Cart