ENAH5802E420 Enamine

1-benzyl-6-bromo-1H-indole

  • MDL number MFCD11894353
  • Cas Number : 481630-30-2
  • Molecular Weight : 286.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 85-88
Iupac Name 1-benzyl-6-bromo-1H-indole
Inchi Code 1S/C15H12BrN/c16-14-7-6-13-8-9-17(15(13)10-14)11-12-4-2-1-3-5-12/h1-10H,11H2
Inchi Key UQGNSLYYFIRZOM-UHFFFAOYSA-N

Description

Melting Point 85-88
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7020689

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7020689
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart