ENAH5802E421 Enamine

5-chloro-4-iodo-1-methyl-1H-pyrazole-3-carboxylic acid

  • Cas Number : 2375262-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7442843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442843
Availability details for
Quantity
  • Availability
  • Add to Cart