ENAH5802E423 Enamine

ethyl 3-bromo-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate

  • Cas Number : 1929549-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7439412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439412
Availability details for
Quantity
  • Availability
  • Add to Cart