ENAH5802E42E Enamine

2-(4-bromo-2,5-dimethoxyphenyl)-2-oxoacetic acid

  • Cas Number : 1511583-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354854
Availability details for
Quantity
  • Availability
  • Add to Cart