ENAH5802E42F Enamine

3-chloro-4-iodoisoquinoline

  • MDL number MFCD31699243
  • Cas Number : 2375269-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7444944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444944
Availability details for
Quantity
  • Availability
  • Add to Cart