ENAH5802E431 Enamine

4-chloro-6-fluoro-2-(methylsulfanyl)pyrimidine

  • MDL number MFCD30615903
  • Cas Number : 2149790-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6415271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6415271
Availability details for
Quantity
  • Availability
  • Add to Cart