ENAH5802E433 Enamine

tert-butyl N-[2-(4-bromo-1H-1,2,3-triazol-1-yl)ethyl]carbamate

  • Cas Number : 2375270-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7440555

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440555
Availability details for
Quantity
  • Availability
  • Add to Cart