ENAH5802E434 Enamine

tert-butyl N-[2-(4-bromo-2H-1,2,3-triazol-2-yl)ethyl]carbamate

  • MDL number MFCD32068806
  • Cas Number : 2375271-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726419
Availability details for
Quantity
  • Availability
  • Add to Cart